Learn More
Medchemexpress LLC Vipivotide tetraxetan | 1702967-37-0 | MFCD31715458 | 99.5% | 1042.14 | C49H71N9O16 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11741050MG
Encompass_Preferred
Vipivotide tetraxetan is a DOTA-conjugated small-molecule ligand that selectively binds prostate-specific membrane antigen (PSMA) and is supplied as a research precursor for radiolabeling with diagnostic and therapeutic radionuclides. It is used in preclinical and clinical research for PSMA-targeted imaging and radioligand therapy.
  • DOTA-conjugated PSMA-targeting ligand suitable for radiolabeling with lutetium-177 or gallium-68.
  • High binding affinity for PSMA enabling targeted imaging and therapy applications.
  • Supplied with high purity (≈99.5%), appropriate for research use.
  • Available in multiple pack sizes, including 50 mg for synthesis and labeling workflows.
  • Useful as a precursor for developing PSMA-targeted radiopharmaceuticals.
Catalog No. 50-002-27522