Learn More
LUNA NANOTECH INC Viral Dna/Rna Purification Kit, 96-Well, 384 Preps
Supplier:  LUNA NANOTECH INC NKP055384
Encompass_Preferred
PuroSPIN-96™ Viral DNA and RNA Purification Kit, 96-Well Plate Format, 384 Preps
PuroSPIN™ Viral DNA/RNA Purification Kit is designed for the extraction of BOTH RNA and DNA from viral sources, such as blood, plasma, serum, milk, buccal or nasal swabs, or saliva. It is intended for viruses such as Coronaviruses, Orthomyxoviruses (Influenza), Adenoviruses, Herpesviruses and others.
This kit greatly simplifies the extraction process by incorporating a convenient easy-to-use extraction technology based on nucleic acid binding silica membranes. The viral particles are digested with an optimized formulation of guanidinium and Proteinase K to disrupt and digest viral capsids and release nucleic acids efficiently from multiple potential sources. Following the lysis step, the nucleic acids are loaded onto silica-based nucleic acid binding membrane and washed to remove protein, salt and other contaminants.  Note that this kit contains a yeast-derived carri
Catalog No. 50-256-6922