Learn More
LUNA NANOTECH INC Viral Dna/Rna Purification Kit, Magnetic, 96 Preps
Supplier:  LUNA NANOTECH INC NKM05596
Encompass_Preferred
PuroMAG™ Viral DNA and RNA Purification Kit, Magnetic, 96 Preps
The PuroMAG™ Viral DNA and RNA Purification Kit is designed to provide a simple and easy-to-use method for extraction of viral genomic nucleic acids from blood, plasma, serum, buccal or nasal swabs, and saliva. This kit can be used for both DNA and RNA viruses as our buffer formulations and column membranes are specially optimized to bind both DNA and RNA nucleic acids. Our kit greatly simplifies the extraction process by incorporating a convenient and easy-to-use extraction technology based on nucleic acid binding to silica-coated magnetic beads.
High quality DNA and RNA extracted with the PuroMAG™ Viral DNA and RNA Purification Kit can be used for a range of downstream applications, such as reverse transcription or qPCR.
This kit offers both low and high throughput applications using a manual magnetic rack or automated purification systems. The high-throughput protoc
Catalog No. 50-256-6923