Learn More
Waters Corp Viridis OBD Prep Column, BEH 2-Ethylpyridine, 5 μm, 130 Å, 19 mm, 150 mm
Supplier:  Waters Corp 186005764
Encompass_Preferred
Designed and tested for Supercritical Fluid Chromatography (SFC), these 5 μm columns demonstrate a wide range of selectivity for greater resolution. The reduction and control of surface silanol activity on Hybrid particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for Supercritical Fluid Chromatography (SFC) applications
  • Provides a wide range of selectivity for enhanced resolution
  • Features reduced and controlled surface silanol activity on Hybrid particles
  • Delivers excellent peak shapes without the need for mobile-phase additives
  • Suitable for well-retained basic achiral compounds
Catalog No. 50-466-248