Learn More
Waters Corp WATERS CORPORATION SFC Preparative Column, 19 x 250 mm, 5 μm, 2-Ethylpyridine
Supplier:  Waters Corp 186004946
Encompass_Preferred
This Viridis Silica 2-Ethylpyridine OBD Prep Column is designed for achiral SFC separations. It provides high reproducibility and predictability due to tight product specifications and low metal content. Available for analytical screening and preparative dimensions, it allows for method optimization and scale-up to Preparative OBD Columns. It offers good retention, peak shape, and selectivity properties.
  • Designed for achiral SFC separations.
  • Ensures high reproducibility and predictability due to tight product specifications and low metal content.
  • Available for analytical screening and preparative dimensions for purification.
  • Separation methods can be optimized and scaled up to Preparative OBD Columns.
  • Provides good retention, peak shape, and selectivity properties with and without additives.
Catalog No. 50-791-253