Learn More
Waters Corp Waters Corporation OBD Prep Column, Silica, 5 μm, 100 Å
Supplier:  Waters Corp 186004924
Encompass_Preferred
These preparative columns combine state-of-the-art media manufacturing with industry-leading column technology, offering enhanced reproducibility for laboratory-scale purification. They are suitable for both analytical and preparative chromatography, with larger formats utilizing patented Optimum Bed Density (OBD) Technology.
  • Offers enhanced reproducibility for laboratory-scale purification.
  • Utilizes patented Optimum Bed Density (OBD) Technology for larger preparative formats.
  • Designed for high reproducibility and predictability.
  • Manufactured with strict product specifications.
  • Features very low metal content.
  • High silanol activity.
Catalog No. 50-791-238
Add to Cart
Request a Quote