Learn More
Waters Corp OBD Prep Column, 100Å, 5 μm, 10 mm X 150 mm
Supplier:  Waters Corp 186008230
Encompass_Preferred
These Supercritical Fluid Chromatography (SFC) columns provide enhanced reproducibility for laboratory purification. They incorporate patented Optimum Bed Density (OBD) Technology for mechanical stability and extended lifetimes, and are manufactured to strict specifications for high reproducibility.
  • State-of-the-art media manufacturing and column technology for high reproducibility in purification.
  • Available for analytical and preparative chromatography.
  • Patented Optimum Bed Density (OBD) Technology enhances mechanical stability and extends column lifetimes.
  • Strict product specifications and minimal metal content ensure high reproducibility and predictability.
  • Designed for improved separation methods and scalability.
  • Ideal for achiral SFC separations, offering good retention, peak shape, and selectivity with or without additives.
Catalog No. 50-239-2799