Learn More
Waters Corp Viridis Silica OBD Prep Column, 100 , 5 m, 10 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186008229
Encompass_Preferred
Viridis Silica OBD Prep Column, 100 , 5 m, 10 mm X 100 mm, 1/pk
Catalog No. 50-239-2578