Learn More
Pharmacal Research Laboratories VIRKON S 50TABLETS 12BTLS/CS
SDP
Supplier:  Pharmacal Research Laboratories 03075F
Encompass
(Potassium Peroxymonosulfate/ Sodium Chloride) is indicated for all your disinfecting and cleaning needs. a broad spectrum disinfectant in an easy to use tablet and is effective against numerous microorganisms affecting animals, including viruses, gram positive and gram-negative bacteria, fungi (molds and yeasts), and Mycoplasma. 50 tablets per bottle and 12 bottles per case. 70693-62-8
Catalog No. NC9549979