Learn More
Cayman Chemical VIrus-URP5BIo-Rad MultIple 1ea
SDP
Supplier:  Cayman Chemical 406981 EA
Encompass_Preferred
Oliogonucleotide recipes are optimized for Bio-Rad CFX96 or CFX384. Includes 4 tubes: • Tube 1: 5X Primers and probes for 4 pathogens and human control • Tube 2: Synthetic positive controls • Tube 3: Spike-in RPP30 control • Tube 4: 2X RT-qPCR enzyme mastermix Organism Type: Virus (-)ssRNA, Virus (+)ssRNA, Multiplexed Reaction Panels: COVID, Viral-URP Colors: Cys5 (RSV A and B), Cys5.5 (Influenza B), HEX (RPP30), TEX615 (SARS-CoV2) Instruments: Bio-Rad CFX96 or CFX384
Catalog No. 50-294-5559