Learn More
Wilmad Labglass Viscometer, Cannon-Fenske. Size: 25
Supplier:  Wilmad Labglass LG11120100
Encompass
Viscometer, Cannon-Fenske. Size: 25; Range Kinematic Centistokes: 0.5-2. Range shown is for an out-flow time of 200 seconds. Tube complies with ASTM D 2515 and is used to obtain kinematic viscosities by ASTM D 445. Certificates of calibration are not supplied with this product.
Catalog No. 50-800-443
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.