Learn More
Wilmad Labglass Viscometer, Cannon-Fenske. Size: 75
Supplier:  Wilmad Labglass LG11120104
Encompass
Viscometer, Cannon-Fenske. Size: 75; Range Kinematic Centistokes: 1.6-8. Range shown is for an out-flow time of 200 seconds. Tube complies with ASTM D 2515 and is used to obtain kinematic viscosities by ASTM D 445. Certificates of calibration are not supplied with this product.
Catalog No. 50-800-445