Learn More
Anton Paar ViscoQC 300 - H a multi-point high viscosity rotational viscometer
Supplier:  Anton Paar 105025
Encompass_Preferred
ViscoQC 300 is a digital rotational viscometer that determines the dynamic viscosity as multiple points for analyzing the flow behavior. Unique benefits include automatic spindle detection by Toolmaster™, automatic guard detection by TruGuard™, unique TruMode™, magnetic coupling for easy one-hand exchange of spindles and digital leveling to ensure the proper alignment of the instrument. It features a data memory and a 7” color glass touchscreen. The instrument is upgradeable with compliance (21 CFR Part 11) and/or additional analysis softwareSpeed range: 0.01 to 250 rpm (freely selectable)Accuracy: ±1.0 % full scale rangeRepeatability: ±0.2 %H model is perfectly suited for high-viscosity samples such as pastes, ointments, gels. Viscosity range: 60* to 320M mPa·s (*with optional double-gap system DG26, with RH1 spindle 800 mPa·s).Includes: Fully assembled instrument, 6 spindles equipped with Toolmaster™, Magnetic coupling, SOP, V-Collect
Catalog No. NC1818578
Add to Cart
Request a Quote