Learn More
Visikol Inc VISIKOL HISTO TISSUE CLEARING
Supplier:  Visikol Inc HISTOM30ML
Encompass_Preferred
Visikol HISTO tissue clearing approach you are now able to rapidly and easily image tissues in 3D without damaging the underlying cellular morphology. The Visikol HISTO approach has been designed to pair with immunolabeling and is reversible so that 3D histology can be followed up with traditional 2D histology. The Visikol HISTO tissue clearing approach for 3D tissue cultures only requires the use of Visikol HISTO-M to render tissues transparent. Visikol HISTO-M has a refractive index of 1.50 and approx. 15 mL is required for a whole 96-well plate of micro-tissues. If you want to test out Visikol HISTO-M before you buy it or have any questions email us at
Catalog No. NC1393516