Learn More
TARGETMOL CHEMICALS INC Vismodegib 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2590200MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Vismodegib (GDC-0449) is a hedgehog pathway inhibitor (IC50 3 nM). It also inhibits P-gp (IC50 3.0 uM), ABCG2 (IC50 1.4 uM). Purity 99.99%
Catalog No. 50-287-7700