Learn More
CTL Scientific Supply Corp VISOCOLOR ECO NICKEL
SDP
Supplier:  CTL Scientific Supply Corp 931040
Encompass_Preferred
VISOCOLOR ECO NICKEL TEST KIT 150 tests/kit. Gradation of scale: 0 - 0.1 - 0.2 - 0.3 - 0.5 - 0.7 - 0.9 - 1.2 - 1.5 ppm Ni2+
Catalog No. 50-300-378