Learn More
TARGETMOL CHEMICALS INC VISOMITIN 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T416825MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Visomitin (SKQ1) is a mitochondria-targeted antioxidant that decreases transmembrane potential and production of reactive oxygen species (ROS). purity: 98%
Catalog No. 50-257-6795