Learn More
ORS Nasco VISOR CLEAR 8 16.5 .040VISOR
Supplier:  ORS Nasco 2804178CL
Encompass_Preferred
Honeywell part# 4178CL. Honeywell Fibre-Metal Visors for F400 Headgear Height: 8 in. Lens Tint: Clear Width: 16.5 in. For Use With: F400 headgear in. Lens Material: Propionate Recommended Applications: Safety
Catalog No. 50-255-1599