Learn More
Medchemexpress LLC VISTA/B7-H5 Mouse 50ug
SDP
Supplier:  Medchemexpress LLC HYP7242850UG
Encompass_Preferred
VISTA/B7-H5 Protein is a type I transmembrane protein expressed primarily in white blood cells that inhibits T cell function VISTA/B7-H5 Protein promotes embryonic stem cell differentiation by inhibiting BMP4 signaling and stimulates MMP14 mediated MMP2 activation VISTA/B7-H5 Protein is highly expressed in tumors VISTA/B7-H5 Protein Mouse (159a a HEK293 His) is the recombinant mouse-derived VISTA/B7-H5 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82920