Learn More
Abcam Vistusertib (AZD2014), 50MG
Supplier:  Abcam AB27362050MG
Encompass_Preferred
MW 462.5 Da, Purity >98%. Novel potent and selective mammalian rapamycin (mTOR) inhibitor (IC₅₀ = 2.8 nM). Highly selective against multiple PI3K isoforms. Also targets P-Akt (IC₅₀ = 80 nM) and pS6 (IC₅₀ = 200 nM). An oral inhibitor with potential anti-neoplastic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1864