Learn More
ELITech Excyte™ Duo-Mix Tube Mixer
SDP
A multifunctional mixer perfect for busy laboratories
Supplier:  ELITech 2900000
Catalog No. 23-024-722
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Compact, quiet mixer is simple to use and designed to perform the mixing of blood samples, controls or other biological liquids
Excyte Duo-Mix - perfect for busy laboratories
  • Accommodates up to 40 samples, plus controls, at one time
  • Fits either 13mm or 8 – 10mm tubes
  • Variety of interchangeable racks optimize workflow
  • Racks can be stored in multiple locations and can be safely transported to the laboratory for mixing
  • Compact and quiet
Product Suggestions
Product Suggestions