Learn More
Medchemexpress LLC Vitamin A Propionate | 7069-42-3 | 98.9% | 342.51 | 100 MG

Supplier: Medchemexpress LLC HYW011180100MG
Vitamin A Propionate is an ester compound of vitamin A. In nutritional studies of foals, different doses have different effects on growth, serum biochemistry, and hematological indicators. Too high or too low doses can produce adverse effects related to the vitamin A concentration in plasma, liver, and kidneys.
- Appearance: Oil
- Color: Colorless to light yellow
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: Pure form -20°C 3 years; In solvent -80°C 6 months, -20°C 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.