Learn More
Medchemexpress LLC Vitamin B12 | 68-19-9 | 99.99% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB031510MM/1ML
Encompass_Preferred
Vitamin B12 (Cyanocobalamin) is a crucial vitamin involved in the metabolism of every cell, particularly affecting DNA synthesis, fatty acid synthesis, and energy production. It plays a key role in brain and nervous system function, and blood formation, offering protection against oxidative stress and benefiting inflammatory diseases.
  • Passes through the blood-brain barrier
  • Involved in cell metabolism
  • Supports DNA synthesis and regulation
  • Aids in fatty acid synthesis and energy production
  • Beneficial for inflammatory diseases
  • Provides protection in oxidative-stress-associated pathologies
Catalog No. 50-004-06773