Learn More
Nanocs Vitamin B12 PEG Biotin, 5000 Da, 5 mg
Supplier:  Nanocs PG2B12BN5K
Encompass
Vitamin B12 PEG Biotin is one of Nanocs bioactive PEG derivatives that can be used for bioassay development. Vitamin B12, also known as cobalamin or cyanocobalamin, is a water soluble vitamin that is involved in the metabolism of cells in human body. Product Specifications:Form: Solid powderColor: RedSolubility: Soluble in waterFunctional ligand: biotinBinding target: Avidin, streptavidin or neutravidinApplications: Bioassay development
Catalog No. NC3450534