Learn More
Cusabio Technology LLC Human Vitamin B12,VB12 ELISA Kit
Supplier:  Cusabio Technology LLC CSBE07903H
Encompass_Preferred
This Human VB12 ELISA Kit was designed for the quantitative measurement of Human VB12 protein in serum, plasma. It is a Competitive ELISA kit, its detection range is 1.56 pg/mL-100 pg/mL and the sensitivity is 1.498 pg/mL. One kit contains 96 wells.
Catalog No. NC2952850