Learn More
LABSYSTEMS DIAGNOSTICS OY VITAMIN C HPLC KIT 200 TESTS
Supplier:  LABSYSTEMS DIAGNOSTICS OY 7250040
Encompass
Vitamin C kit is used in routine clinical laboratories for the quantitative determination ofvitamin C in serum and plasma to detect deficiencies or hypervitaminosis. The separation of the vitamins and their specific internal standards is performed on a specific analytical column and all the analytes are measured by UV detection.
Catalog No. NC2865595