Learn More
ABclonal Technology Vitamin D 25-OH-VD Rabbit mAb
SDP
Supplier:  ABclonal Technology A25579100UL
Encompass_Preferred
25-OH-VD is the primary circulating form of vitamin D in human body after its initial metabolism It serves as the preferred biomarker for assessing vitamin D status due to its plasma concentration accurately reflecting total body stores Derived from the hydroxylation of vitamin D precursors (vitamin D3 or D2) in the liver 25-OH-VD is a precursor to the biologically active 1 25-(OH)2-D3 formed in the kidneys Its long half-life and stable levels make it the standard analyte for laboratory tests to diagnose vitamin D deficiency or excess crucial in preventing and treating conditions like osteoporosis rickets and various chronic diseases linked to vitamin D including cardiovascular issues immune disorders muscle dysfunction diabetes and certain cancers
Catalog No. 50-000-10960