Learn More
U.S. Pharmacopeia Cholecalciferol, 67-97-0, MFCD00078131, 5amp, 30mg/ampule
Supplier:  U.S. Pharmacopeia 1131009
Encompass
Molecular formula C27H44O, Molecular weight 384.64, Melting Point 183.2 - 192.2 °F (84 - 89 °C), Synonyms: Vitamin D3, Colecalciferol, Activated 7-Dehydrocholesterol
Catalog No. NC9859279