Learn More
Medchemexpress LLC Vitamin D3-d7 | 1627523-19-6 | C27H37D7O | 500 UG
SDP
Supplier:  Medchemexpress LLC HY15398S500UG
Encompass_Preferred
Vitamin D3-d7 is a deuterium-labeled form of Vitamin D3 (Cholecalciferol), a naturally occurring vitamin known to induce cell differentiation and prevent the proliferation of cancer cells. It incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, primarily as tracers for quantitation during drug development, with potential to influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-76327