Learn More
ATCC Vitamin Supplement for Bacteriological Culture Media, Package of 5 (10 mL/vial) (ATCC MD-VS™)
Supplier:  ATCC MDVS
Encompass
Vitamin Supplement (ATCC MD-VS™) is a sterile additive for bacteriological culture media, especially for extremophile cultivation. Based on Wolfe’s vitamin solution, it includes folic acid, biotin, thiamine, B12, and other essential nutrients. Supplied as 5 vials (10 mL each).
Catalog No. 50-238-4130
 Restricted Item