Learn More
Cayman Chemical Vitellogenin rainbow trout E
SDP
Supplier:  Cayman Chemical 10004994480 WELL
Encompass_Preferred
The Vtg (rainbow trout) ELISA is a double-antibody immunometric (sandwich) ELISA for analyzing Vtg in plasma samples.
Catalog No. 50-288-7076