Learn More
Cayman Chemical VItelogenInAtlntIc salmon 10ug
SDP
Supplier:  Cayman Chemical 47014410 UG
Encompass_Preferred
The lyophilized Atlantic salmon Vtg may be used as a positive control in immunoblotting. Freshly reconstituted Vtg may also be used as a standard in a quantitative ELISA. This product contains bovine serum albumin which will appear as a strong protein band in acrylamide gels.
Catalog No. 50-294-5280