Learn More
Medchemexpress LLC Vitexin-2"-O-rhamnoside | 64820-99-1 | 578.52 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN05341MG
Encompass_Preferred
Vitexin-2"-O-rhamnoside is an orally active flavonoid glycoside. It inhibits apoptosis and promotes cytokine secretion, demonstrating significant activity against DNA synthesis in MCF-7 cells. This compound enhances immune function, improves the absorption of active compounds, and exhibits antioxidant properties, making it valuable for cardiovascular and immune-related research.
  • Orally active flavonoid glycoside
  • Inhibits apoptosis and caspase-3 activity
  • Increases PI3K/Akt phosphorylation levels
  • Promotes cytokine (IL-2, IL-6, IL-12) secretion
  • Strongly inhibits DNA synthesis in MCF-7 cells (IC50 of 17.5 μM)
  • Enhances immune function and improves absorption of active compounds
  • Possesses antioxidant activity
  • Useful in cardiovascular and immune-related disease studies
Catalog No. 50-003-88853