Learn More
Medchemexpress LLC Vitexin-2''-O-rhamnoside | 64820-99-1 | 578.52 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN053410MM/1ML
Encompass_Preferred
Vitexin-2"-O-rhamnoside is an orally active flavonoid glycoside that plays a role in various biological processes. It inhibits apoptosis, increases phosphorylation levels of PI3K/Akt, and inhibits caspase-3 and SOD activity. The compound also promotes the secretion of cytokines such as IL-2, IL-6, and IL-12. It strongly inhibits DNA synthesis in MCF-7 cells with an IC50 of 17.5 μM. This product is used in the study of cardiovascular disease and immune-related diseases.
  • Enhances immune function
  • Improves absorption of active compounds
  • Exhibits antioxidant activity
  • Utilized in research for cardiovascular and immune-related conditions
Catalog No. 50-003-88965