Learn More
Medchemexpress LLC Vitexin-2"-O-rhamnoside | 64820-99-1 | MFCD29913069 | 578.52 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN05345MG
Encompass_Preferred
Vitexin-2"-O-rhamnoside is an orally active flavonoid glycoside that inhibits Apoptosis, increases the phosphorylation levels of PI3K/Akt, inhibits caspase-3, SOD activity, and promotes cytokine (IL-2, IL-6, and IL-12) secretion. It strongly inhibits DNA synthesis in MCF-7 cells with an IC50 of 17.5 μM, enhances immune function, improves the absorption of active compounds, and has antioxidant activity. It is used in the study of cardiovascular disease and immune-related diseases.
  • Inhibits apoptosis.
  • Increases phosphorylation levels of PI3K/Akt.
  • Inhibits caspase-3 and SOD activity.
  • Promotes cytokine (IL-2, IL-6, and IL-12) secretion.
  • Strongly inhibits DNA synthesis in MCF-7 cells with an IC50 of 17.5 μM.
  • Enhances immune function.
  • Improves absorption of active compounds.
  • Has antioxidant activity.
  • Used in the study of cardiovascular disease and immune-related diseases.
Catalog No. 50-003-88455