Learn More
Sheldon Manufacturing Inc VITON DOOR GASKET
SDP
Supplier:  Sheldon Manufacturing Inc 3450671
Encompass_Preferred
Vacuum Oven Gasket 18"x18" Viton composition for 4.5 cuft vacuum ovens
Catalog No. NC0886918