Learn More
STEMCELL Technologies Vitronectin XF™, Size: 20 mL
SDP
Supplier:  STEMCELL Technologies 1000763
Encompass
Defined, xeno-free matrix that supports the growth and differentiation of human pluripotent stem cells under serum-free, feeder-free conditions. 10 x 2 mL vials.
Catalog No. NC1989226
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov