Learn More
Medchemexpress LLC VK-28 | 312611-92-0 | MFCD06254693 | 97.2% | 287.36 | C16H21N3O2 | 5 MG

Supplier: Medchemexpress LLC HYW6862165MG
VK-28 is a brain-permeable iron chelator used as a research tool in neurodegeneration studies. It inhibits basal and Fe/ascorbate-induced mitochondrial membrane lipid peroxidation (IC50 = 12.7 μM) and has shown neuroprotective effects in ICV-6-OHDA models. The compound is characterized by molecular weight 287.36, formula C16H21N3O2, CAS 312611-92-0, and is supplied at high purity for laboratory research.
- Brain-permeable iron chelator that inhibits mitochondrial membrane lipid peroxidation.
- Demonstrated IC50 of 12.7 μM for lipid peroxidation inhibition.
- Shown neuroprotective effects in ICV-6-OHDA experimental models.
- High purity suitable for research use (97.17%).
- Molecular weight 287.36 and formula C16H21N3O2 for reference.
- Storage guidance: powder at -20°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.