Learn More
Medchemexpress LLC VKGILS-NH2 | 942413-05-0 | 99.3% | 614.78 | 10 MG

Supplier: Medchemexpress LLC HYP131010MG
VKGILS-NH2 is a reversed amino acid sequence control peptide for SLIGKV-NH2, a protease-activated receptor 2 (PAR2) agonist. It has no effect on DNA synthesis in cells.
- Reversed amino acid sequence control peptide
- No effect on DNA synthesis in cells
- Solid, white to off-white appearance
- Sequence: Val-Lys-Gly-Ile-Leu-Ser-NH2
- Target: PAR2
- Storage as powder: -80°C for 2 years, -20°C for 1 year
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.