Learn More
Medchemexpress LLC VKGILS-NH2 | 942413-05-0 | 99.3% | 614.78 | 5 MG

Supplier: Medchemexpress LLC HYP13105MG
VKGILS-NH2 is a control peptide for SLIGKV-NH2, a protease-activated receptor 2 (PAR2) agonist. This peptide has no effect on DNA synthesis in cells.
- Reversed amino acid sequence
- No effect on DNA synthesis in cells
- Acts as a PAR agonist
- Appears as a white to off-white solid
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.