Learn More
Medchemexpress LLC HY-111663 10mg Medchemexpress, VL285 CAS:1448188-57-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11166310MG
Encompass_Preferred
Medchemexpress, HY-111663 10mg VL285 CAS:1448188-57-5 VL285 is a potent VHL ligand with an IC50 of 0.34 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8940