Learn More
Medchemexpress LLC VL285 | 1448188-57-5 | 99.3% | 532.65 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11166310MM/1ML
Encompass_Preferred
VL285 is a potent VHL ligand with an IC50 of 0.34 μM.
  • Potent VHL ligand.
  • Attenuates the ability of HaloPROTAC3 to induce the degradation of GFP-HaloTag7.
  • Useful in cancer research.
  • Applicable in cancer targeted therapy.
  • Used as a ligand for E3 ligase.
Catalog No. 50-004-39880