Learn More
Medchemexpress LLC VL285 | 1448188-57-5 | 99.3% | 532.65 | 25 MG

Supplier: Medchemexpress LLC 25MGHY111663
VL285 is a potent VHL ligand with an IC50 of 0.34 μM. It is for research use only and is not intended for patient use. Treatment with VL285 attenuates the ability of HaloPROTAC3 to induce the degradation of GFP-HaloTag7.
- Potent VHL ligand
- IC50 of 0.34 μM
- Attenuates degradation of GFP-HaloTag7 by HaloPROTAC3
- For research use only
- Molecular weight of 532.65
- Purity of 99.3%
- Soluble in DMSO (100 mg/mL)
- Available as a white to off-white solid
- Stable as powder at -20°C for 3 years, 4°C for 2 years
- Stable in solvent at -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.