Learn More
Selleck Chemical LLC VL285 S0095-25MG
SDP
Supplier:  Selleck Chemical LLC S009525MG
Encompass_Preferred
VL285 is a potent VHL ligand that degrades HaloTag7 fusion proteins Cotreatment with excess VL285 the core VHL ligand from which HaloPROTAC3 is derived is able to significantly reduce HaloPROTAC3 mediated activity to 50% degradation
Catalog No. 50-000-40291