Learn More
ACROBiosystems Fluorescent VLP isotype control (VLP-NF2P4) is expressed from human 293 cells (HEK293).
Supplier:  ACROBiosystems VLPNF2P430UG
Encompass_Preferred
Virus-like particles (VLPs) are highly structured protein particles composed of single or multiple structural proteins of the virus. Its morphological structure is similar to that of natural virus particles, does not contain virus accounting, and has strong immunogenicity and biological activity. The empty shell structure of most VLP is icosahedral or helical structure, which is composed of several specific structural proteins. At present, it is widely used in vaccine production and antigen presentation.
Catalog No. 50-225-3936