Learn More
Chemglass Life Sciences Valve, Chem-Vac, Chem-Cap, Hi-Vac, Low Hold-Up, 0-8mm Bore
SDP
Supplier:  Chemglass Life Sciences CG95502
Encompass_Preferred
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Low Hold-Up, 0-8mm Bore. Similar to CG-953 CHEM-VAC valve but with the arms bent to minimize hold up of fluids. See CG-961 for replacement CHEM-CAP plug and CG-905 for glass portion only.
Catalog No. CG95502