Learn More
Medchemexpress LLC Vm24-toxin (Vaejovis mexicanus peptide 24) | 1373890-79-9 | 3863.59 | 1 MG
SDP
Catalog No. 5000399214
Encompass_Preferred
Vm24-toxin (Vaejovis mexicanus peptide 24) is a 36-residue peptide that acts as a potent and selective Kv1.3 blocker with a Kd of approximately 3 pM in lymphocytes. It exhibits over 1500-fold higher affinity for Kv1.3 compared to other assayed potassium channels. Vm24-toxin possesses a distorted cystine-stabilized α/β motif, comprising a single-turn α-helix and a three-stranded antiparallel β-sheet, stabilized by four disulfide bridges. This peptide can attenuate the CD4+ effector memory T cell response to T cell receptor (TCR) stimulation.
  • Potent and selective Kv1.3 blocker
  • High affinity for Kv1.3 over other potassium channels (>1500-fold)
  • Attenuates CD4+ effector memory T cell response to T cell receptor (TCR) stimulation
Catalog No. 50-003-99214 Supplier Medchemexpress LLC Supplier No. HYP5917A1MG
Vm24-toxin (Vaejovis mexicanus peptide 24) is a 36-residue peptide that acts as a potent and selective Kv1.3 blocker with a Kd of approximately 3 pM in lymphocytes. It exhibits over 1500-fold higher affinity for Kv1.3 compared to other assayed potassium channels. Vm24-toxin possesses a distorted cystine-stabilized α/β motif, comprising a single-turn α-helix and a three-stranded antiparallel β-sheet, stabilized by four disulfide bridges. This peptide can attenuate the CD4+ effector memory T cell response to T cell receptor (TCR) stimulation.
  • Potent and selective Kv1.3 blocker
  • High affinity for Kv1.3 over other potassium channels (>1500-fold)
  • Attenuates CD4+ effector memory T cell response to T cell receptor (TCR) stimulation