Learn More
Medchemexpress LLC VMO1 Human HEK293 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7728050UG
Encompass_Preferred
Vitelline membrane outer layer protein 1 homolog (VMO1) is first characterized in the outer layer of the vitelline membrane of hen s eggs where VMO1is present together with lysozyme VMO2 and ovomucin VMO1 is a secreted protein and exerts important functions in inner ear and tear film VMO1 may also interact with glycosylated proteins VMO1 Protein Human (HEK293) is the recombinant human-derived VMO1 protein expressed by HEK293 with tag free
Catalog No. 50-001-86523