Learn More
Cayman Chemical VnP16trIfluoroacetAT salt 5mg
SDP
Supplier:  Cayman Chemical 388395 MG
Encompass_Preferred
A synthetic peptide fragment of vitronectin; induces attachment and cell spreading of isolated human osteogenic cells when applied to cell plates at 9.1 UG/cm2; induces osteogenesis of human skin-derived mesenchymal precursors and MC3T3-E1 mouse calvarial osteoblast precursors; decreases IL-1α-induced bone loss and decreases the number of calvarial osteoclasts in mice; increases bone mineral density and decreases trabecular bone loss in a mouse model of ovariectomy-induced bone loss
Catalog No. 50-291-9034