Learn More
Selleck Chemical LLC VO-Ohpic trihydrate 10mg 476310-60-8
SDP
Supplier:  Selleck Chemical LLC S817410MG
Encompass_Preferred
VO-Ohpic is a potent inhibitor of PTEN (phosphatase and tensin homolog) with IC50 of 35 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5745